missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beschreibung
The Human IgG Heavy chain antibody from Proteintech is a mouse monoclonal antibody generated with nature protein of Human IgG Heavy chain. This antibody recognizes human antigen. The Human IgG Heavy chain antibody has been validated for the following applications: WB, IHC, IF-P, ELISA analysis.
Spezifikation
Spezifikation
| Antigen | IgG Heavy chain |
| Anwendungen | Western Blot, Immunohistochemistry, Immunofluorescence, ELISA |
| Klon | 1C6B4 |
| Konzentration | 1000 μg/mL |
| Konjugat | Unconjugated |
| Wirtsspezies | Mouse |
| Immunogen | nature protein |
| Reinigungsverfahren | Protein A purification |
| Menge | 150 μL |
| Primär oder sekundär | Primary |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?