missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human IgG kappa Recombinant Mouse Monoclonal Antibody (SAA0278)
Mouse Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA554604
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Compatible with Direct ELISA.
Human IgG, also known as human immunoglobulin G, is an antibody that is produced by the immune system in response to antigens. It is the most abundant immunoglobulin in the blood and plays a key role in defense against pathogens. IgG works by binding to specific antigens and promoting their clearance from the body through various mechanisms, including phagocytosis and complement activation. IgG is also involved in other immune responses, such as antibody-dependent cellular cytotoxicity and neutralization of toxins. Human IgG can be used in research to study the immune response to pathogens and to develop diagnostic tests and therapeutic interventions for infectious and autoimmune diseases.
Spezifikation
| Human IgG kappa | |
| Recombinant Monoclonal | |
| 1.66 mg/mL | |
| PBS with no preservative | |
| Mouse | |
| Protein A/G | |
| RUO | |
| Human | |
| Antibody | |
| VHH-mFc |
| ELISA | |
| SAA0278 | |
| Unconjugated | |
| Immunoglobulin G; ImmunoglobulinG | |
| Recombinant Human IgG Kappa Chain. | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur