Learn More
Human IL-1 RI Antibody, R&D Systems™
Beschreibung
IL-1 RI Polyclonal specifically detects IL-1 RI in Human samples. It is validated for Western Blot, Neutralization.
Spezifikation
Spezifikation
| Antigen | IL-1 RI |
| Anwendungen | Western Blot, Neutralization |
| Klassifikation | Polyclonal |
| Konzentration | LYOPH |
| Konjugat | Unconjugated |
| Verdünnung | Western Blot 1 ug/mL, Neutralization 5-10 ug/mL |
| Gen-Alias | CD121 antigen-like family member A, CD121a, CD121a antigen, IL1R, IL1R1, IL1RI, IL-1RI, IL1RT1, IL-1RT1, IL-1RT-1, Interleukin 1 Receptor 1, interleukin 1 receptor, type I, interleukin receptor 1, interleukin-1 receptor type 1, p80 |
| Gensymbole | IL1R1 |
| Wirtsspezies | Goat |
| Immunogen | S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-1 RI |
| Mehr anzeigen |