missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-15R alpha Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 14 publications
Marke: R&D Systems AF247-SP
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
IL-15R alpha Polyclonal specifically detects IL-15R alpha in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Neutralization, Immunocytochemistry, CyTOF-ready.
Spezifikation
| IL-15R alpha | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD215, CD215 antigen, IL15 R alpha, IL-15 R alpha, IL-15 receptor subunit alpha, IL15RA, IL-15RA, IL-15R-alpha, interleukin 15 receptor, alpha, interleukin-15 receptor subunit alpha, MGC104179 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3601 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Neutralization, Immunocytochemistry, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, Neutralization 1-4 ug/mL, Immunocytochemistry 1-15 ug/mL, CyTOF-ready | |
| EAW86418 | |
| IL15RA | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-15 R alpha Ile31-Thr172 Accession # EAW86418 | |
| 25 μg | |
| Primary | |
| Detects human IL-15 R alpha in direct ELISAs and Western blots. In direct ELISAs, less than 10% cross-reactivity with recombinant human IL-2 R gamma is observed. | |
| Human | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur