missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-17E/IL-25 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
221.00 € - 2060.00 €
Spezifikation
| Antigen | IL-17E/IL-25 |
|---|---|
| Klon | 182203 |
| Konzentration | LYOPH |
| Verdünnung | Western Blot 1 ug/mL, ELISA, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready |
| Anwendungen | Western Blot, ELISA, Flow Cytometry, CyTOF |
| Product Code | Brand | Menge | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Menge | Price | Quantity & Availability | |||||
18755719
![]() |
R&D Systems
MAB1258-SP |
25 μg |
221.00 €
25 Mikrogramm |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | |||||
18760577
![]() |
R&D Systems
MAB1258-100 |
100 μg |
528.00 €
100 Mikrogramm |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | |||||
18740948
![]() |
R&D Systems
MAB1258-500 |
500 μg |
2060.00 €
500 Mikrogramm |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | |||||
Description
IL-17E/IL-25 Monoclonal specifically detects IL-17E/IL-25 in Human samples. It is validated for Western Blot, ELISA, Intracellular Staining by Flow Cytometry, CyTOF-ready.Specifications
| IL-17E/IL-25 | |
| LYOPH | |
| Western Blot, ELISA, Flow Cytometry, CyTOF | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| IL17E, IL-17E, IL25, IL-25, interleukin 25, Interleukin-17E, interleukin-25 | |
| IL25 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 182203 | |
| Western Blot 1 ug/mL, ELISA, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Human | |
| Q9H293 | |
| 64806 | |
| E. coli-derived recombinant human IL-17E Tyr33-Gly177 Accession # Q9H293 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human IL-17E in direct ELISAs and Western blots. Shows 100% cross-reactivity with recombinant mouse IL-17E and no cross-reactivity with recombinant human (rh) IL-17, rhIL-17B, rhIL-17C, or rhIL-17F. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title