missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human IL-4, Animal-Free Recombinant Protein, PeproTech®
Click to view available options
Menge:
250 μg
5 μg
50 μg
Packungsgröße:
250 Mikrogramm
5 Mikrogramm
50 Mikrogramm
Beschreibung
Recombinant Human IL-4 is a 15.1 kDa globular protein containing 130 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Interleukin-4 (IL-4) is a pleiotropic, immune-modulatory cytokine produced by activated T cells, and serves as a ligand for interleukin 4 receptor. IL-4 has been found to have the broadest range of actions. The interleukin 4 receptor also binds to IL13, which may contribute to many overlapping functions of this cytokine and IL13. STAT6, a signal transducer and activator of transcription, has been shown to play an important role in mediating the immune regulatory signal of IL-4. The IL-4 gene, IL-3, IL-5, IL-13, and CSF2 form a cytokine gene cluster on chromosome 5q. In particular, IL-4 is located in close proximity to IL13 on the chromosome. Further, the IL-4 gene, IL13 and IL5 are found to be coordinated by several long-range regulatory elements over a 120 kilobase range on the chromosome. IL-4 delta 2 is a known splice variant of IL-4.
Spezifikation
Spezifikation
| Zugriffsnummer | P05112 |
| Zur Verwendung mit (Anwendung) | Functional Assay |
| Format | Lyophilized |
| Zusammensetzung | protein with no preservative |
| Gen-ID (Entrez) | 3565 |
| Molekulargewicht | 15.1 kDa |
| Name | Human IL-4, Animal-Free |
| Reinigungsverfahren | HPLC, SDS-PAGE |
| Menge | 250 μg |
| Kennzeichnung | RUO |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur