missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-5 R alpha/CD125 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
Marke: R&D Systems AF-253-SP
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
IL-5R alpha/CD125 Polyclonal specifically detects IL-5R alpha/CD125 in Human samples. It is validated for Western Blot, Flow Cytometry, Neutralization, CyTOF-ready.
Spezifikation
| IL-5R alpha/CD125 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| IL5RA | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-5 R alpha/CD125 | |
| 25 μg | |
| Primary | |
| Detects human IL-5 R alpha in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human (rh) IL-1 RII, rhIL-2 R beta , rhIL-2 R gamma , rhIL-3 R is observed and less than 1% cross-reactivity with rhIL-1 RI, rhIL-4 R, rhIL-6 R, rhIL-7 R, rhIL-9 R, and rhIL-10 R is observed. | |
| Human | |
| IgG |
| Western Blot, Flow Cytometry, Neutralization, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Neutralization 1-3 ug/mL, CyTOF-ready | |
| CD125, CD125 antigen, CDw125CD125, HSIL5R3, IL-5 receptor subunit alpha, IL5R, IL5R alpha, IL-5R subunit alpha, IL5RA, IL-5RA, IL-5R-alpha, interleukin 5 receptor type 3, interleukin 5 receptor, alpha, interleukin-5 receptor alpha chain, interleukin-5 receptor alpha subunit, interleukin-5 receptor subunit alpha, MGC26560 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3568 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur