missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-5 R alpha/CD125 Antibody, R&D Systems™

Mouse Monoclonal Antibody
247.00 € - 580.00 €
Spezifikation
| Antigen | IL-5R alpha/CD125 |
|---|---|
| Klon | 1036601 |
| Verdünnung | Flow Cytometry 0.25 ug/10^6 cells |
| Anwendungen | Flow Cytometry |
| Klassifikation | Monoclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
18704577
![]() |
R&D Systems
MAB2531-SP |
25 μg |
247.00 €
25 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
18761337
![]() |
R&D Systems
MAB2531-100 |
100 μg |
580.00 €
100 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
IL-5R alpha/CD125 Monoclonal antibody specifically detects IL-5R alpha/CD125 in Human samples. It is validated for Flow CytometrySpezifikation
| IL-5R alpha/CD125 | |
| Flow Cytometry 0.25 ug/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| 3568 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
| 1036601 | |
| Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Human | |
| CD125 antigen, CDw125CD125, HSIL5R3, IL-5 receptor subunit alpha, IL5R, IL-5R subunit alpha, IL-5RA, IL-5R-alpha, interleukin 5 receptor type 3, interleukin 5 receptor, alpha, interleukin-5 receptor alpha chain, interleukin-5 receptor alpha subunit, interleukin-5 receptor subunit alpha, MGC26560 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-5 R alpha/CD125, Asp21-Arg335, Accession # O08665 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title