missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Integrin alpha 1/CD49a Antibody, R&D Systems™

Sheep Polyclonal Antibody
Marke: R&D Systems AF5676
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Integrin alpha 1/CD49a Polyclonal specifically detects Integrin alpha 1/CD49a in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready.
Spezifikation
| Integrin alpha 1/CD49a | |
| Polyclonal | |
| Unconjugated | |
| P56199 | |
| ITGA1 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Integrin alpha 1/CD49a Phe29-Pro1141 Accession # P56199 | |
| 100 μg | |
| Primary | |
| Detects human Integrin alpha 1/CD49a in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) Integrin alpha 2, rhIntegrin alpha 11, recombinant mouse (rm) Integrin alpha 2, and rmIntegrin alpha 11 is observed. | |
| Human | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF | |
| LYOPH | |
| Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, CyTOF-ready | |
| CD49 antigen-like family member A, CD49a, CD49a antigen, integrin, alpha 1, ITGA1, Laminin and collagen receptor, very late activation protein 1, VLA-1, VLA1integrin alpha-1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 3672 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur