missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Integrin alpha 4 beta 7/LPAM-1 (Research Grade Vedolizumab Biosimilar) Alexa Fluor™ 350-conjugated Antibody, R&D Systems™

Human Monoclonal Antibody
Marke: R&D Systems FAB10078U-100UG
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Integrin alpha 4 beta 7/LPAM-1 Monoclonal specifically detects Integrin alpha 4 beta 7/LPAM-1 in Human samples. It is validated for Flow Cytometry.
Spezifikation
| Integrin alpha 4 beta 7/LPAM-1 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| Human | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects human Integrin alpha 4 beta 7/LPAM-1 in direct ELISAs. This non-therapeutic antibody uses the same variable region sequence as the therapeutic antibody Vedolizumab. This product is for research use only. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG1 |
| Flow Cytometry | |
| Hu117 | |
| Flow Cytometry 0.25-1 μg/106 cells | |
| 269C wild type, CD49 antigen-like family member D, CD49d, CD49d antigen, CD49d/beta7, CD49Dantigen CD49D, alpha-4 subunit of VLA-4 receptor, IA4, integrin alpha 4, integrin alpha-4, integrin alpha-4 subunit, Integrin alpha-IV, integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor), LPAM1, LPAM-1, MGC90518, very late activation protein 4 receptor, alpha 4 subunit, VLA-4 subunit alpha | |
| Human Integrin alpha 4 beta 7/LPAM-1 | |
| 100 μg | |
| 3676 | |
| Human | |
| Supernatant |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur