missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Isocitrate Dehydrogenase 1/IDH1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marke: R&D Systems MAB7049-SP
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Isocitrate Dehydrogenase 1/IDH1 Monoclonal specifically detects Isocitrate Dehydrogenase 1/IDH1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Knockout Validated.
Spezifikation
| Isocitrate Dehydrogenase 1/IDH1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.25 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunocytochemistry 8-25 ug/mL, Knockout Validated | |
| O75874 | |
| IDH1 | |
| E. coli-derived recombinant human Isocitrate Dehydrogenase 1/IDH1 Ser2-Leu414 Accession # O75874 | |
| 25 μg | |
| Primary | |
| Detects human Isocitrate Dehydrogenase 1/IDH1 in ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, KnockDown | |
| 843219 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Cytosolic NADP-isocitrate dehydrogenase, EC 1.1.1.42, IDCD, IDH, IDH1, IDP, IDPC, isocitrate dehydrogenase [NADP] cytoplasmic, isocitrate dehydrogenase 1 (NADP+), soluble, NADP(+)-specific ICDH, NADP-dependent isocitrate dehydrogenase, cytosolic, NADP-dependent isocitrate dehydrogenase, peroxisomal, Oxalosuccinate decarboxylase, PICD | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3417 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur