missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LC3B Antibody, R&D Systems™
Mouse Monoclonal Antibody
228.00 € - 492.00 €
Spezifikation
| Antigen | LC3B |
|---|---|
| Klon | 1082854 |
| Verdünnung | Western Blot 2 μg/mL |
| Anwendungen | Western Blot |
| Klassifikation | Monoclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
30279902
|
R&D Systems
MAB11606-SP |
25 μg |
228.00 €
25 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
30279918
|
R&D Systems
MAB11606-100 |
100 μg |
492.00 €
100 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
LC3B Monoclonal antibody specifically detects LC3B in Human samples. It is validated for Western BlotSpezifikation
| LC3B | |
| Western Blot 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
| Apg8b, ATG8F, Autophagy-related protein LC3 B, Autophagy-related ubiquitin-like modifier LC3 B, LC3B, lc3b autophagy marker, lc3-i, ii, LC3II, MAP1 light chain 3-like protein 2, MAP1A/1BLC3, MAP1A/MAP1B LC3 B, map1lc3b, MAP1LC3B-a, microtubule associated protein 1 light chain 3 beta, microtubule associated protein 3 b, microtubule associated protein 3 beta, microtubule-associated proteins 1A/1B light chain 3, microtubule-associated proteins 1A/1B light chain 3B | |
| Synthetic Peptide, Accession # Q9GZQ8 | |
| Primary | |
| Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. |
| 1082854 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9GZQ8 | |
| 81631 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20°C to -70 °C under sterile conditions after reconstitution. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts