missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LILRB4/CD85k/ILT3 Antibody, R&D Systems™
Description
ILT3/CD85k/LILRB4 Polyclonal specifically detects ILT3/CD85k/LILRB4 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
Specifications
| Antigen | ILT3/CD85k/LILRB4 |
| Anwendungen | Western Blot, Flow Cytometry, CyTOF |
| Klassifikation | Polyclonal |
| Konzentration | LYOPH |
| Konjugat | Unconjugated |
| Verdünnung | Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready |
| Gen-Zugriffsnummer | ABM86208 |
| Gen-Alias | CD85 antigen-like family member K, CD85k, CD85k antigen, HM18, ILT3, ILT-3, ILT3CD85K, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR5, LIR5LILRB5, LIR-5subfamily B, member 4, member 4 |
| Gensymbole | LILRB4 |
| Wirtsspezies | Goat |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?