missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LILRB4/CD85k/ILT3 APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marke: R&D Systems FAB24251A-100
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
ILT3/CD85k/LILRB4 Monoclonal specifically detects ILT3/CD85k/LILRB4 in Human samples. It is validated for Flow Cytometry.
Spezifikation
| ILT3/CD85k/LILRB4 | |
| Monoclonal | |
| APC | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD85 antigen-like family member K, CD85k, CD85k antigen, HM18, ILT3, ILT-3, ILT3CD85K, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR5, LIR5LILRB5, LIR-5subfamily B, member 4, member 4 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 11006 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 293623 | |
| Flow Cytometry 10 uL/10^6 cells | |
| ABM86208 | |
| LILRB4 | |
| Mouse myeloma cell line NS0-derived recombinant human ILT3/CD85k Pro17-His257 Accession # ABM86208 | |
| 100 Tests | |
| Primary | |
| Detects human ILT3/CD85k in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with with recombinant human (rh) ILT5 is observed. No cross-reactivity with rhILT1, rhILT2, rhILT4, rhILT7, rhILT11, rhLIR-6 or rhLIR-8 is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2a |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur