Learn More
Human Lipocalin-2/NGAL Antibody, R&D Systems™
GREENER_CHOICE
Artikelnummer. 18721647
Change view
Click to view available options
Menge:
100 μg
25 μg
500 μg
Packungsgröße:
100 Mikrogramm
25 Mikrogramm
500 Mikrogramm
Artikelnummer. Menge unitSize
18721647 25 μg 25 Mikrogramm
18752939 100 μg 100 Mikrogramm
18730028 500 μg 500 Mikrogramm
3 options
Artikelnummer. 18721647
Marke: R&D Systems MAB1757SP
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Rat Monoclonal Antibody has been used in 9 publications
Lipocalin-2/NGAL Monoclonal specifically detects Lipocalin-2/NGAL in Human samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen Lipocalin-2/NGAL
Anwendungen Western Blot, Immunohistochemistry
Klassifikation Monoclonal
Klon 220310
Konzentration LYOPH
Konjugat Unconjugated
Verdünnung Western Blot 0.2-1 ug/mL, Immunohistochemistry 8-25 ug/mL
Zusammensetzung Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gen-Zugriffsnummer P80188
Gen-Alias 24p3, 25 kDa alpha-2-microglobulin-related subunit of MMP-9, HNL, LCN2, lipocalin 2, lipocalin 2 (oncogene 24p3), Lipocalin2, Lipocalin-2, migration-stimulating factor inhibitor, MSFI, neutrophil gelatinase-associated lipocalin, NGAL, NGALlipocalin-2, Oncogene 24p3, p25, Siderocalin, Uterocalin
Gensymbole LCN2
Wirtsspezies Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Lipocalin-2/NGAL Gln21-Gly198 Accession # P80188
Reinigungsverfahren Protein A or G purified from hybridoma culture supernatant
Menge 25 μg
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 3934
Testspezifität Detects human Lipocalin-2/NGAL in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human (rh) Lipocalin-1 or rmLipocalin-2. This antibody also recognizes human Lipocalin-2/MMP-9 complexes in Western blots under non-reducing conditions.
Rekonstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Zielspezies Human
Inhalt und Lagerung Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a