Learn More
Human LOX-1/OLR1 Alexa Fluor™ 647-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Marke:  R&D Systems FAB1798R-100UG
Artikelnummer. 18747667
  • 551.00 € / 100 Mikrogramm
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
LOX-1/OLR1 Monoclonal specifically detects LOX-1/OLR1 in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
LOX-1/OLR1
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
P78380
OLR1
Mouse myeloma cell line NS0-derived recombinant human LOX-1/OLR1 Ser61-Gln273 Accession # P78380
RUO
4973
Human
IgG2b
Flow Cytometry
331212
Alexa Fluor 647
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
CLEC8A, CLEC8ASLOX1, C-type lectin domain family 8 member A, hLOX-1, Lectin-like oxidized LDL receptor 1, Lectin-like oxLDL receptor 1, Lectin-type oxidized LDL receptor 1, LOX1, LOX-1, LOX1ox LDL receptor 1, LOXIN, OLR1, oxidised low density lipoprotein (lectin-like) receptor 1, oxidized low density lipoprotein (lectin-like) receptor 1, oxidized low-density lipoprotein receptor 1, oxidized low-density lipoprotein receptor 1, soluble form, Ox-LDL receptor 1, SCARE1, scavenger receptor class E, member 1, SR-E1
Mouse
100 μg
Primary
Detects human LOX-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant mouse LOX-1.
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.
For Research Use Only (RUO)