missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human LPO (aa 199-294) Control Fragment Recombinant Protein
Click to view available options
Menge:
100 μl
Unit Size:
100 Mikroliter
Description
Highest antigen sequence indentity to the following orthologs: Mouse (84%), Rat (84%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-83026 (PA5-83026. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
This gene encodes an oxidoreductase secreted from salivary, mammary, and other mucosal glands that functions as a natural antibacterial agent. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
| Zugriffsnummer | P22079 |
| Konzentration | ≥5.0 mg/mL |
| Zur Verwendung mit (Anwendung) | Blocking Assay, Control |
| Zusammensetzung | 1 M urea, PBS with no preservative; pH 7.4 |
| Gen-ID (Entrez) | 4025 |
| Name | Human LPO (aa 199-294) Control Fragment |
| Menge | 100 μl |
| Kennzeichnung | RUO |
| Gen-Alias | 5830499B15Rik; airway lactoperoxidase; EC 1.11.17 antibody; lactoperoxidase; lactoperoxydase; Lpo; LPO antibody; MGC129990; MGC129991; Salivary peroxidase; SAPX; SAPX antibody; SPO; SPO antibody |
| Gebräuchliche Bezeichnung | LPO |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction