missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Lysyl Oxidase Homolog 2/LOXL2 Antibody, R&D Systems™
Goat Polyclonal Antibody
Marke: R&D Systems AF2639
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Lysyl Oxidase Homolog 2/LOXL2 Polyclonal specifically detects Lysyl Oxidase Homolog 2/LOXL2 in Human samples. It is validated for Western Blot, Simple Western, Immunoprecipitation.
Spezifikation
| Lysyl Oxidase Homolog 2/LOXL2 | |
| Polyclonal | |
| Unconjugated | |
| AAH00594 | |
| LOXL2 | |
| Mouse myeloma cell line NS0-derived recombinant human Lysyl Oxidase Homolog 2/LOXL2 Gln26-Gln774 Accession # AAH00594 | |
| 100 μg | |
| Primary | |
| Detects human Lysyl Oxidase Homolog 2/LOXL2 in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot, Immunoprecipitation | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunoprecipitation 25 ug/mL | |
| EC 1.4.3, EC 1.4.3.-, LOL2, LOR2, LOXL2, lysyl oxidase homolog 2, lysyl oxidase related 2, lysyl oxidase-like 2, Lysyl oxidase-like protein 2, Lysyl oxidase-related protein 2, Lysyl oxidase-related protein WS9-14, WS9-14 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4017 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur