missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Amphiphysin/AMPH Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB65271
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Amphiphysin/AMPH Monoclonal specifically detects Amphiphysin/AMPH in Human, Mouse samples. It is validated for Western Blot.
Specifikationer
| Amphiphysin/AMPH | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| P49418 | |
| AMPH | |
| E. coli-derived recombinant human Amphiphysin/AMPH Asn204-Gly300 Accession # P49418 | |
| 100 μg | |
| Primary | |
| Detects human Amphiphysin/AMPH in direct ELISAs. | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| 694121 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AMPH, AMPH1, amphiphysin, amphiphysin (Stiff-Man syndrome with breast cancer 128kDa autoantigen), amphiphysin (Stiff-Mann syndrome with breast cancer 128kD autoantigen), amphiphysin I | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 273 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering