missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse FABP4/A-FABP Antibody, R&D Systems™
Rat Monoclonal Antibody
Marke: R&D Systems MAB3150
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
FABP4/A-FABP Monoclonal specifically detects FABP4/A-FABP in Human, Mouse samples. It is validated for Western Blot, ELISA.
Spezifikation
| FABP4/A-FABP | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, ELISA | |
| P15090 | |
| FABP4 | |
| E. coli-derived recombinant human FABP4/A-FABP Met1-Ala132 Accession # P15090 | |
| 100 μg | |
| Primary | |
| Detects human FABP4/A-FABP in direct ELISAs and human and mouse FABP4/A-FABP in Western blots. In direct ELISAs, 100% cross-reactivity with recombinant mouse (rm) FABP4, 50% cross-reactivity with recombinant human (rh) FABP3, 15% cross-reactivity with rhFABP3, and no cross-reactivity with rhFABP8 or rhFABP9 is observed. | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA | |
| 804914 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Adipocyte lipid-binding protein, Adipocyte-type fatty acid-binding protein, AFABP, A-FABP, A-FABPAFABP, ALBP, aP2, fatty acid binding protein 4, adipocyte, Fatty acid-binding protein 4, fatty acid-binding protein, adipocyte | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2167 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur