Learn More
Human/Mouse/Rat SOX2 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 119 publications
Marke:  R&D Systems AF2018
Product Code. 18081962
  • 618.00 € / 100 Mikrogramm
om dit product te kopen Registreer vandaag om een webaccount aan te maken
This item is not returnable. View return policy
Description
Description
SOX2 Polyclonal specifically detects SOX2 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation (ChIP), Immunocytochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SOX2
Polyclonal
Western Blot 1 ug/mL, Simple Western 5 ug/mL, Immunohistochemistry 3-25 ug/mL, Chromatin Immunoprecipitation (ChIP) 5 ug/5 x 10^6 cells, Immunocytochemistry 5-15 ug/mL
ANOP3, MCOPS3, MGC2413, SRY (sex determining region Y)-box 2, SRY-related HMG-box gene 2, transcription factor SOX2, transcription factor SOX-2
Goat
Affinity Purified
RUO
6657
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Western Blot, Immunohistochemistry, ChIP Assay, Immunocytochemistry
Unconjugated
P48431
SOX2
E. coli-derived recombinant human SOX2 Gly135-Met317 Accession # P48431
100 μg
Primary
Detects human SOX2 in direct ELISAs and Western blots.
Human, Mouse, Rat
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications