Learn More
Human NFIL3/E4BP4 Antibody, Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Marke: R&D Systems MAB11657-100
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
NFIL3 Monoclonal antibody specifically detects NFIL3 in Human samples. It is validated for Western Blot,Simple Western
Spezifikation
| NFIL3 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2μm filtered solution in PBS. | |
| E4BP4, E4BP4IL3BP1, IL3BP1, Interleukin-3 promoter transcriptional activator, Interleukin-3-binding protein 1, NF-IL3A, NFIL3AE4 promoter-binding protein 4, nuclear factor interleukin 3 regulated protein, nuclear factor interleukin-3-regulated protein, nuclear factor, interleukin 3 regulated, Transcriptional activator NF-IL3A | |
| E. coli- derived recombinant human NFIL-3, Lys140-Gly462, Accession # Q16649 | |
| 100 μg | |
| Primary | |
| Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| Purified |
| Western Blot, Simple Western | |
| 1095812 | |
| Western Blot 2 μg/mL, Simple Western 20 μg/mL | |
| Q16649 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4783 | |
| Human | |
| Antibody | |
| IgG2a |