missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NGL-1/LRRC4C Antibody, R&D Systems™

Description
NGL-1/LRRC4C Monoclonal specifically detects NGL-1/LRRC4C in Human samples. It is validated for Immunohistochemistry.
Specifications
Specifications
| Antigen | NGL-1/LRRC4C |
| Anwendungen | Immunohistochemistry |
| Klassifikation | Monoclonal |
| Klon | 755720 |
| Konzentration | LYOPH |
| Konjugat | Unconjugated |
| Verdünnung | Immunohistochemistry 8-25 ug/mL |
| Zusammensetzung | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
| Gen-Zugriffsnummer | Q9HCJ2 |
| Gen-Alias | KIAA1580NGL-1NGL1Netrin-G1 ligand, leucine rich repeat containing 4C, leucine-rich repeat-containing protein 4C, LRRC4C, NGL1 |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?