Learn More
Human NKG2D/CD314 PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Artikelnummer. 18706407
Change view
Click to view available options
Menge:
100 Tests
25 Tests
Packungsgröße:
100 Tests
25 Tests
Artikelnummer. Menge unitSize
18706407 25 Tests 25 Tests
18749716 100 Tests 100 Tests
2 options
Artikelnummer. 18706407
Marke: R&D Systems FAB139P025
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Mouse Monoclonal Antibody has been used in 26 publications
NKG2D/CD314 Monoclonal specifically detects NKG2D/CD314 in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen NKG2D/CD314
Anwendungen Flow Cytometry
Klassifikation Monoclonal
Klon 149810
Konjugat R-PE
Verdünnung Flow Cytometry 10 uL/10^6 cells
Zusammensetzung Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gen-Alias CD314, CD314 antigen, D12S2489E, FLJ17759, FLJ75772, Killer cell lectin-like receptor subfamily K member 1, killer cell lectin-like receptor subfamily K, member 1, KLR, KLRK1, NK cell receptor D, NKG2-D, NKG2-D type II integral membrane protein, NKG2-D-activating NK receptor, NKG2DDNA segment on chromosome 12 (unique) 2489 expressed sequence
Gensymbole KLRK1
Wirtsspezies Mouse
Immunogen BaF3 mouse pro-B cell line transfected with human NKG2D/CD314
Reinigungsverfahren Protein A or G purified from hybridoma culture supernatant
Menge 25 Tests
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 22914
Testspezifität Detects human NKG2D/CD314 in Western blots.
Zielspezies Human
Inhalt und Lagerung Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG1