Learn More
Human NKp30/NCR3 PE-conjugated Antibody, R&D Systems™
Beschreibung
NKp30/NCR3 Monoclonal specifically detects NKp30/NCR3 in Human samples. It is validated for Flow Cytometry.
Spezifikation
Spezifikation
| Antigen | NKp30/NCR3 |
| Anwendungen | Flow Cytometry |
| Klassifikation | Monoclonal |
| Klon | 210845 |
| Konjugat | R-PE |
| Verdünnung | Flow Cytometry 10 uL/10^6 cells |
| Zusammensetzung | Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide |
| Gen-Alias | Activating natural killer receptor p30, CD337, CD337 antigen, CD337activating NK-A1 receptor, LY117,1C7NK-p30, lymphocyte antigen 117, natural cytotoxicity triggering receptor 3, Natural killer cell p30-related protein, NCR3, NKP30, NKp30MALS |
| Gensymbole | NCR3 |
| Wirtsspezies | Mouse |
| Mehr anzeigen |