missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Nociceptin/Orphanin FQ ELISA Kit

Artikelnummer. 30829955
Ansicht ändern
Click to view available options
Menge:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge
30829955 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 30829955 Lieferant Invitrogen™ Lieferanten-Nr. EEL175
Black Friday beginnt hier
Verwenden Sie den Aktionscode 27984
Alle Werbeaktionen anzeigen
 Eingeschränkter Artikel
Verfügbar ab: 11-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

ELISA

The Human Orphanin FQ/Nociceptin (OFQ/N) ELISA quantitates OFQ/N in serum, plasma and other biological fluids. Principle of the method The Human Orphanin FQ/Nociceptin (OFQ/N) solid phase sandwich ELISA (enzyme linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target specific antibody has been pre coated in the wells of the supplied microplate. Samples are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the binding of the second (detector) antibody to the target on a different epitope from the capture antibody. An antibody conjugated with enzyme binds the formed sandwich. After incubation and washing steps to rid the microplate of unbound substances. Rigorous validation: Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation. For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

Nociceptin/orphanin FQ is a neuropeptide that acts as a ligand for the nociceptin receptor. It modulates pain perception, stress responses, and various physiological processes. Structurally, it is a 17-amino acid peptide derived from the prepronociceptin protein with the sequence FGGFTGARKSARKLANQ. Nociceptin/orphanin FQ is widely distributed in the central nervous system and peripheral tissues, indicating its involvement in diverse functions.
TRUSTED_SUSTAINABILITY

Spezifikation

Assay-Bereich 4.69-300 pg/mL
Assay-Empfindlichkeit: 2.81 pg/mL
Konjugat HRP
Immunassay-Kit-Format Sandwich ELISA Kit
Produkttyp ELISA
Probentyp Cell Lysates, Plasma, Serum, Tissue Homogenates
Zielspezies Human
Zur Verwendung mit (Geräte) Absorbance Microplate Reader
Gen-Alias N/OFQ; OFQ/NN/OFQ; OFQ/N;
Interassay-VK <10%
Intraassay-VK <10%
Kitinhalt Pre-coated 96 well plate, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer
Marker oder Farbstoff Biotin
Menge 96 Tests
Kennzeichnung RUO
Probenvolumen Cell Lysate 100 μL, Plasma 100 μL, Serum 100 μL, Tissue Homogenate 100 μL
Versandbedingung Wet or Dry Ice
Lagerungsbedingungen Refer to product documentation for component specific storage temperature.
Ziel Nociceptin/Orphanin FQ
Prüfzeit 1 hr 20 min
Gesamte Assay-Zeit 3 hr 30 min
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.