missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human P-Rex1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marke: R&D Systems MAB8076
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
P-Rex1 Monoclonal specifically detects P-Rex1 in Human samples. It is validated for Western Blot, Simple Western.
Spezifikation
| P-Rex1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Simple Western 5 ug/mL | |
| Q8TCU6 | |
| PREX1 | |
| E. coli-derived recombinant human P-Rex1 Lys1313-Arg1434 Accession # Q8TCU6 | |
| 100 μg | |
| Primary | |
| Detects human P-Rex1 in ELISAs and Western Blots. | |
| Human | |
| Purified |
| Western Blot | |
| 869704 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| KIAA1415, phosphatidylinositol 34,5-trisphosphate-dependent Rac exchanger 1 protein, phosphatidylinositol-34,5-trisphosphate-dependent Rac exchange factor 1, PRex1, ptdIns(34,5)-dependent Rac exchanger 1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 57580 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur