missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Plectin (aa 3341-3427) Control Fragment Recombinant Protein
Click to view available options
Menge:
100 μl
Packungsgröße:
100 Mikroliter
Beschreibung
Highest antigen sequence indentity to the following orthologs: Mouse (87%), Rat (87%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-55280 (PA5-55280. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Plectin is a prominent member of an important family of structurally and in part functionally related proteins, termed plakins or cytolinkers, that are capable of interlinking different elements of the cytoskeleton. Plakins, with their multi-domain structure and enormous size, not only play crucial roles in maintaining cell and tissue integrity and orchestrating dynamic changes in cytoarchitecture and cell shape, but also serve as scaffolding platforms for the assembly, positioning, and regulation of signaling complexes. Plectin is expressed as several protein isoforms in a wide range of cell types and tissues from a single gene located on chromosome 8 in humans. The expression of specific plectin isoforms was found to be dependent on cell type (tissue) and stage of development and it appears that each cell type (tissue) contains a unique set (proportion and composition) of plectin isoforms which carry out distinct and specific functions.
Spezifikation
Spezifikation
| Zugriffsnummer | Q15149 |
| Konzentration | ≥5.0 mg/mL |
| Zur Verwendung mit (Anwendung) | Blocking Assay, Control |
| Zusammensetzung | 1 M urea, PBS with no preservative; pH 7.4 |
| Gen-ID (Entrez) | 5339 |
| Name | Human Plectin (aa 3341-3427) Control Fragment |
| Menge | 100 μl |
| Kennzeichnung | RUO |
| Gen-Alias | AA591047; AU042537; EBS1; EBSMD; EBSND; EBSO; EBSOG; EBSPA; HD1; Hemidesmosomal protein 1; LGMD2Q; PCN; Plec; PLEC1; PLEC1b; Plectin; plectin 1; plectin 1, intermediate filament binding protein 500 kDa; plectin-1; Plectin-6; Pltn |
| Gebräuchliche Bezeichnung | Plectin |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur