Learn More
Human PTP beta/zeta/PTPRZ Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB2688-SP
This item is not returnable.
View return policy
Description
PTP beta/zeta/PTPRZ Monoclonal specifically detects PTP beta/zeta/PTPRZ in Human samples. It is validated for Western Blot.
Specifications
| PTP beta/zeta/PTPRZ | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P23471 | |
| PTPRZ1 | |
| E. coli-derived recombinant human PTP beta/zeta/PTPRZ Gly302-Asn754 Accession # P23471 | |
| 25 μg | |
| Primary | |
| Detects human PTP beta/zeta/PTPRZ in Western blots. | |
| Human | |
| Purified |
| Western Blot | |
| 279244 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 3F8 CSPG, HPTPZ, phosphacan, protein tyrosine phosphatase, receptor-type, Z polypeptide 1, PTP18, PTPRZ, PTPRZ1, PTPZ, receptor-type, zeta polypeptide 1, RPTPB, RPTPbeta | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5803 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |