Learn More
STEMCELL Technologies Human Recombinant FGF-9
Beschreibung
- Fibroblast growth factor (FGF) 9 is a pleiotropic member of the FGF family
- FGFs are heparin-dependent regulators of cell proliferation and differentiation (Itoh & Ornitz)
- FGF-9 signals by binding FGFR3 (IIIb) and the IIIc splice variants of the FGFR1, FGFR2, and FGFR3 receptors (Santos-Ocampo et al.; Mohammadi et al.; Plotnikov et al.) In mice, deletion of FGF-9 is lethal and expression of FGF-9 affects development of the skeleton, cerebellum, lungs, heart, vascular system, digestive tract, and testes (Colvin et al.; Harada et al.; Lin et al.) Altered levels of FGF-9 have also been reported in various types of human cancers (Leushacke et al.; Abdel-Rahman et al.)
Spezifikation
Spezifikation
| Name | Fibroblast growth factor-9, GAF, Glia-activating factor, HBGF-9 |
| Menge | 50 μg |
| Zelltyp | Airway Cells; Mesenchymal Cells, PSC-Derived; Mesenchymal Stem and Progenitor Cells |
| Reinheits- oder Qualitätsgrad | ≥95% |
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?