Learn More
STEMCELL Technologies Human Recombinant Follistatin
Beschreibung
- Follistatin, a glycosylated monomeric protein, is a modulator of transforming growth factor beta (TGF-β) superfamily signaling
- It binds to and inhibits the function of activin, myostatin, growth differentiation factors, and bone morphogenetic proteins (BMP) (Hansen & Plomgaard)
- Follistatin inhibits mesoderm induction, suppresses synthesis and secretion of pituitary follicle-stimulating hormone, regulates liver regeneration, and causes infertility (Guo et al.; Iemura et al.) Follistatin exhibits anti-inflammatory effects, and it could be used as a biomarker in cancer (Hansen & Plomgaard)
Spezifikation
Spezifikation
| Name | Activin-binding protein, FS, FSH-suppressing protein, FSP |
| Menge | 100 μg |
| Zelltyp | Mesoderm, PSC-Derived |
| Reinheits- oder Qualitätsgrad | ≥95% |