Learn More
STEMCELL Technologies Human Recombinant TNF-beta
Artikelnummer. p-7217650
Encompass_Preferred
Change view
Click to view available options
Menge:
10 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge
17149621 10 μg
17159621 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17149621 Lieferant STEMCELL Technologies Lieferanten-Nr. 78125
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Tumor necrosis factor beta
  • Tumor necrosis factor-beta (TNF-β), also known as lymphotoxin-alpha, is a member of the TNF family
  • It is a pro-inflammatory cytokine that activates NF-kB, MAPK, and PI3K/AKT pathways upon binding to TNF receptors 1 and 2
  • It is produced mainly by T cells, though other cells can express TNF-β at lower levels (Chu)
  • TNF-β is involved in autoimmune disorders, lymph node development, and mediating the inflammatory demyelination process (Seleznik et al.; McCarthy et al.)
Spezifikation
Name LT-α, Lymphotoxin-alpha, TNF-beta, TNFSF1, Tumor necrosis factor-β
Menge 10 μg
Zelltyp T Cells
Reinheits- oder Qualitätsgrad ≥95%