missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Rictor Antibody, R&D Systems™
Rat Monoclonal Antibody
Marke: R&D Systems MAB4598
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Rictor Monoclonal specifically detects Rictor in Human samples. It is validated for Western Blot.
Spezifikation
| Rictor | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q6R327 | |
| RICTOR | |
| E. coli-derived recombinant human Rictor Met1-Leu250 Accession # Q6R327 | |
| 100 μg | |
| Primary | |
| Detects endogenous human Rictor in Western blots. | |
| Human | |
| Purified |
| Western Blot | |
| 454313 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AVO3, AVO3 homolog, hAVO3, KIAA1999DKFZp686B11164, mAVO3, MGC39830, PIA, pianissimo, rapamycin-insensitive companion of mTOR, RPTOR independent companion of MTOR, complex 2, TORC2-specific protein AVO3 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 253260 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur