missing translation for 'onlineSavingsMsg'
Få mere at vide

Human ROR alpha/NR1F1 PE-conjugated Antibody, R&D Systems™

Artikelnummer. 18125001
Skift visning
Klik for at se tilgængelige muligheder
Menge:
100 Tests
25 Tests
Pakningsstørrelse:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
18125001 100 Tests 100 Tests
18742657 25 Tests 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 18125001 Leverandør R&D Systems Leverandørnr. IC8924P100

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Mouse Monoclonal Antibody has been used in 1 publication

ROR alpha/NR1F1 Monoclonal specifically detects ROR alpha/NR1F1 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen ROR alpha/NR1F1
Anwendungen Flow Cytometry
Klassifikation Monoclonal
Klon 784652
Konjugat R-PE
Verdünnung Intracellular Staining by Flow Cytometry 10 uL/10^6 cells
Zusammensetzung Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gen-Zugriffsnummer P35398
Gen-Alias DKFZp686M2414, MGC119326, NR1F1, NR1F1MGC119329, nuclear receptor ROR-alpha, Nuclear receptor RZR-alpha, Nuclear receptor subfamily 1 group F member 1, RAR-related orphan receptor A, RAR-related orphan receptor alpha, retinoic acid receptor-related orphan receptor alpha, retinoid-related orphan receptor alpha, Retinoid-related orphan receptor-alpha, ROR1, ROR2, ROR3, RORA, RZRA, RZR-ALPHA, RZRAROR-alpha, transcription factor RZR-alpha
Gensymbole RORA
Wirtsspezies Mouse
Immunogen E. coli-derived recombinant human ROR alpha/NR1F1 Arg47-His163 Accession No. P35398
Reinigungsverfahren Protein A or G purified from hybridoma culture supernatant
Menge 100 Tests
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 6095
Testspezifität Detects human ROR alpha/NR1F1 in direct ELISAs.
Zielspezies Human
Inhalt und Lagerung Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2b
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.