missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Serum Amyloid A1/A2 Antibody, R&D Systems™

Description
Serum Amyloid A1/A2 Monoclonal specifically detects Serum Amyloid A1/A2 in Human samples. It is validated for ELISA.
Specifications
Specifications
| Antigen | Serum Amyloid A1/A2 |
| Anwendungen | ELISA |
| Klassifikation | Monoclonal |
| Klon | 924912 |
| Konjugat | Unconjugated |
| Verdünnung | ELISA |
| Zusammensetzung | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. |
| Gen-Zugriffsnummer | P0DJI8 |
| Wirtsspezies | Mouse |
| Immunogen | E. coli-derived recombinant human Serum Amyloid A1 Arg19-Tyr122 Accession # P0DJI8 |
| Show More |
RUO
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?