missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human IL-7R alpha/CD127 (soluble) Fc Recombinant Protein, PeproTech®

Artikelnummer. 17994162
Change view
Click to view available options
Menge:
1 mg
10 μg
100 μg
250 μg
50 μg
500 μg
Packungsgröße:
1 Milligramm
10 Mikrogramm
100 Mikrogramm
250 Mikrogramm
50 Mikrogramm
500 Mikrogramm
Artikelnummer. Menge unitSize
17994162 10 μg 10 Mikrogramm
17944192 50 μg 50 Mikrogramm
17984312 500 μg 500 Mikrogramm
17934192 250 μg 250 Mikrogramm
17914362 1 mg 1 Milligramm
17944362 100 μg 100 Mikrogramm
6 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 17994162

Marke: Gibco™ 20007RA10UG

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Recombinant Protein

Recombinant Human sIL-7 Receptor alpha Fc is a 51.3 kDa protein containing 452 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

CD127 (Interleukin-7, IL-7) is a glycoprotein involved in the regulation of lymphopoiesis. The response of cells to CD127 is dependent on the presence of the interleukin 7 receptor (IL7R); the active receptor is an alpha/gamma chain heterodimer. CD127 consists of an alpha chain and a gamma chain. The gamma(c) chain, which also associates with the interleukin-2 receptor, serves primarily to activate signal transduction by the IL7R complex, while the alpha chain of IL7R determines specific signaling events through its association with cytoplasmic signaling molecules. CD127 promotes the proliferation of precursor B cells, thymocytes, T cell progenitors, and mature CD4+ and CD8+ T cells. The biological effects of IL7 are mediated by the binding of IL7 to the specific cell surface receptor. Diseases associated with CD127 dysfunction include severe combined immunodeficiency and T cell negative/B cell negative/NK positive severe combined immunodeficiency.
TRUSTED_SUSTAINABILITY

Spezifikation

Zugriffsnummer P16871
Zur Verwendung mit (Anwendung) Functional Assay
Format Lyophilized
Zusammensetzung protein with no preservative
Gen-ID (Entrez) 3575
Molekulargewicht 51.3 kDa
Name Human IL-7 R alpha/CD127 (soluble) Fc
Menge 10 μg
Kennzeichnung RUO
Endotoxin-Konzentration <1 EU/ μg
Gen-Alias CD127; CD127 antigen; CDW127; IL 7 receptor; IL 7 R a; IL 7 R subunit alpha; IL 7 R a; IL 7 Ra; IL 7 Ra; IL7 receptor; IL-7 receptor alpha chain; IL7 receptor subunit alpha; IL-7 receptor subunit alpha; IL7R; IL-7 R; IL7R a; IL7R alpha; IL7R subunit alpha; IL-7 R subunit alpha; IL7R a; IL7RA; IL-7 RA; IL-7 Ralpha; IL-7 R-alpha; IL7Ra; ILRA; interleukin 7 receptor; interleukin 7 receptor alpha chain; interleukin 7 receptor isoform H5-6; Interleukin7 receptor subunit alpha; interleukin-7 receptor subunit alpha; MGC107557; sCD127; soluble IL 7 receptor; soluble IL 7 R
Gebräuchliche Bezeichnung CD127
Biologische Aktivität Determined by its ability to bind human IL-7 in. a functional ELISA.
Konjugat Unconjugated
Rekombinant Recombinant
Proteinmarkierung Fc-tag
Sequenz ESGYAQNGDL EDAELDDYSF SCYSQLEVNG SQHSLTCAFE DPDVNITNLE FEICGALVEV KCLNFRKLQE IYFIETKKFL LIGKSNICVK VGEKSLTCKK IDLTTIVKPE APFDLSVVYR EGANDFVVTF NTSHLQKKYV KVLMHDVAYR QEKDENKWTH VNLSSTKLTL LQRKLQPAAM YEIKVRSIPD HYFKGFWSEW SPSYYFRTPE INNSSGEMDG GPKSCDKTHT CPPCPAPELL GGPSVFLFPP KPKTTLMISR TPEVTCVVVD VSHEDPEVKF NWYVDGVEVH NAKTKPREEQ YNSTYRVVSV LTVLHQDWLN GKEYKCKVSN KALPAPIEKT ISKAKGQPRE PQVYTLPPSR DELTKNQVSL TCLVKGFYPS DIAVEWESNG QPENNYKTTP PVDLSDGSFF LYSKLTVDKS RWQQGNVFSC SVMHEALHNH YTQKSLSLSP GK
Inhalt und Lagerung -20°C
Expressionssystem CHO cells
Form Lyophilized
Reinheits- oder Qualitätsgrad ≥ 95% by SDS-PAGE gel and HPLC analyses.
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.