missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SLC1A5 Antibody, R&D Systems™
Description
SLC1A5 Monoclonal antibody specifically detects SLC1A5 in Human samples. It is validated for Immunohistochemistry,Immunocytochemistry
Specifications
Specifications
| Antigen | SLC1A5 |
| Anwendungen | Immunohistochemistry, Immunocytochemistry |
| Klassifikation | Monoclonal |
| Klon | 3085B |
| Konjugat | Unconjugated |
| Verdünnung | Immunohistochemistry 1-10 μg/mL, Immunocytochemistry 1-10 μg/mL |
| Zusammensetzung | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gen-Zugriffsnummer | Q9JKD8 |
| Gen-Alias | AAAT, Alanine/serine/cysteine transporter 2, ASC amino acid transporter-2, ASCT2, ATB(0), ATBO, baboon M7 virus receptor, M7V1, M7VS1, neutral amino acid transporter B, neutral amino acid transporter B(0), R16, RD114 virus receptor, RD114/simian type D retrovirus receptor, RDR, RDRC, Sodium-dependent neutral amino acid transporter type 2, solute carrier family 1 (neutral amino acid transporter), member 5, Solute carrier family 1 member 5 |
| Wirtsspezies | Rabbit |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?