missing translation for 'onlineSavingsMsg'
Learn More

Human SOX17 Antibody, R&D Systems™
GREENER_CHOICE

Artikelnummer. 18733878
Change view
Click to view available options
Menge:
25 μg
Packungsgröße:
25 Mikrogramm
Artikelnummer. Menge unitSize
18733878 25 μg 25 Mikrogramm
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 18733878

Marke: R&D Systems MAB1924SP

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Mouse Monoclonal Antibody has been used in 22 publications

SOX17 Monoclonal specifically detects SOX17 in Human samples. It is validated for Immunocytochemistry.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen SOX17
Anwendungen Immunocytochemistry
Klassifikation Monoclonal
Klon 245013
Konzentration LYOPH
Konjugat Unconjugated
Verdünnung Immunocytochemistry 8-25 ug/mL
Zusammensetzung Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gen-Zugriffsnummer Q9H6I2
Gen-Alias FLJ22252, SRY (sex determining region Y)-box 17, SRY-related HMG-box transcription factor SOX17, transcription factor SOX-17, VUR3
Gensymbole SOX17
Wirtsspezies Mouse
Immunogen E. coli-derived recombinant human SOX17 Asp177-Val414 Accession # Q9H6I2
Reinigungsverfahren Protein A or G purified from hybridoma culture supernatant
Menge 25 μg
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 64321
Testspezifität Detects human SOX17 in direct ELISAs. In direct ELISAs, this antibody does not cross-react with recombinant human SOX2, 3, 7, or 10.
Rekonstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Zielspezies Human
Inhalt und Lagerung Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG3
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.