Learn More
Human SR-AI/MSR1 Antibody, R&D Systems™
GREENER_CHOICE
Artikelnummer. 18737158
Change view
Click to view available options
Menge:
100 μg
25 μg
Packungsgröße:
100 Mikrogramm
25 Mikrogramm
Artikelnummer. Menge unitSize
18737158 25 μg 25 Mikrogramm
18129020 100 μg 100 Mikrogramm
2 options
Artikelnummer. 18737158
Marke: R&D Systems MAB27081SP
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Mouse Monoclonal Antibody has been used in 3 publications
SR-AI/MSR Monoclonal specifically detects SR-AI/MSR in Human samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction.
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen SR-AI/MSR
Anwendungen Western Blot, Blockade of Receptor-ligand Interaction
Klassifikation Monoclonal
Klon 351620
Konzentration LYOPH
Konjugat Unconjugated
Verdünnung Western Blot 1 ug/mL, Blockade of Receptor-ligand Interaction
Zusammensetzung Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gen-Zugriffsnummer P21757
Gen-Alias CD204, CD204 antigen, Macrophage acetylated LDL receptor I and II, macrophage scavenger receptor 1, macrophage scavenger receptor type III, MSR1, phSR1, phSR2, SCARA1, SCARA1macrophage scavenger receptor types I and II, Scavenger receptor class A member 1, scavenger receptor class A, member 1, SR-A, SRAI
Gensymbole MSR1
Wirtsspezies Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human SR-AI Lys77-Leu451 Accession # P21757
Reinigungsverfahren Protein A or G purified from hybridoma culture supernatant
Menge 25 μg
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 4481
Testspezifität Detects human SR-AI in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant mouse SR-AI.
Rekonstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Zielspezies Human
Inhalt und Lagerung Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Mostrar más Mostrar menos
For Research Use Only
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.