missing translation for 'onlineSavingsMsg'
Learn More

Human TGF beta 1, Invitrogen™

Artikelnummer. 15112079
Click to view available options
Menge:
5 μg
Packungsgröße:
5 Mikrogramm
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 15112079

Marke: Invitrogen 14834862

om dit product te kopen Registreer vandaag om een webaccount aan te maken

Dieser Artikel wurde eingestellt und ist leider nicht mehr verfügbar. Sehen Sie sich das Produkt an, um alternative Produktvorschläge zu sehen oder kontaktieren Sie unseren Technischen Support unter 056 618 41 11.
Alternative Produkte anzeigen

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Recombinant Protein

TGF beta-1 (TGFB1, Transforming Growth Factor Beta 1) is a polypeptide member of the transforming growth factor beta superfamily of cytokines, found almost ubiquitously in tissues. Transforming growth factor (TGF)-b is stored in the extracellular matrix as a latent complex with its pro-domain. Activation of TGF beta-1 requires the binding of aV integrin to an RGD sequence in the prodomain and exertion of force on this domain, which is held in the extracellular matrix by latent TGF-b binding proteins. Latent forms are complexes of TGF beta-1, an amino-terminal portion of the TGF-beta precursor, designated TGF-LAP (TGF-latency associated peptide), and a specific binding protein, known as LTBP. TGF beta-1 helps regulates proliferation, differentiation, adhesion, migration in many cell types. Many cells have TGF beta receptors, and the protein positively and negatively regulates many other growth factors. TGF beta-1 is cleaved into a latency-associated peptide and a mature TGF beta-1 peptide, and is found in either a latent form composed of a TGFB1 homodimer, a LAP homodimer, and a latent TGFB1-binding protein, or in an active form composed of a TGF beta-1 homodimer. The mature peptide may also form heterodimers with other TGF beta family members. The gene for TGF beta-1 is frequently upregulated in tumor cells, and mutations in this gene result in Camurati-Engelmann disease and cystic fibrosis. Description: Human TGF beta 1 (Transforming Growth Factor beta 1) is a member of a superfamily of homologous, disulfide-linked, homodimeric proteins that regulate the proliferation and differentiation of normal and transformed cells. Human TGF beta 1 is a 25 kDa protein with each subunit containing 112 aa residues. Applications Reported: Recombinant human TGF beta1 is biologically active and can inhibit IL-4 induced proliferation of mouse HT-2 cells. Recombinant human TGF beta1 is useful as an ELISA standard. Applications Tested: The ED50 of this protein, as measured by inhibition of IL-4 induced proliferation in HT-2 cells, is less than or equal to 40 pg/mL. This corresponds to a specific activity of greater than or equal to 2.5 x 10e7 Units/mg. Source: CHO cells expressed amino acids Ala279-Ser390; accession &Num;BC001180. Bioactivity: The ED50 of this protein, as measured by inhibition of IL-4 induced proliferation in HT-2 cells, is less than or equal to 40 pg/mL. This corresponds to a specific activity of greater than or equal to 2.5 x 10e7 Units/mg. Endotoxin: Less than 0.01ng/ug cytokine as determined by the LAL assay. Purity: >98% as determined by SDS-PAGE. Molecular Weight: Mature TGFb1 polypeptide has a predicted molecular mass of 12,794. On reducing SDS-PAGE the protein migrates as a 13 kDa polypeptide and on non-reducing SDS page the cystine-linked homodimer migrates as a 25 kDa protein. Storage and handling: For best recovery, quick-spin vial prior to opening. Use in a sterile environment. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2um post-manufacturing filtered.
TRUSTED_SUSTAINABILITY

Spezifikation

Zugriffsnummer P01137
Konzentration 0.1mg/mL
Zur Verwendung mit (Anwendung) Control, ELISA, Functional Assay, Neutralization
Zusammensetzung protein with 0.1M glycine, 1% BSA and no preservative; pH 3.0
Gen-ID (Entrez) 7040
Molekulargewicht Mature TGFb1 polypeptide has a predicted molecular mass of 12,794. On reducing SDS-PAGE the protein migrates as a 13kDa polypeptide and on non-reducing SDS page the cystine-linked homodimer migrates as a 25kDa protein.
Name Human TGF beta 1
pH-Bereich 3.0
Reinigungsverfahren SDS-PAGE
Menge 5 μg
Quelle CHO Cells
Lagerungsbedingungen -80°C
Kennzeichnung RUO
Strukturform CHO cells expressed amino acids ala279 - ser390; accession #BC001180.
Endotoxin-Konzentration Less than 0.01ng/μg cytokine as determined by the LAL assay.
Gebräuchliche Bezeichnung Human TGF beta 1
Gensymbol TGFB1
Biologische Aktivität The ED50 of this protein, as measured by inhibition of IL-4 induced proliferation in HT-2 cells, is less than or equal to 40pg/mL. This corresponds to a specific activity of greater than or equal to 2.5 x 107 Units/mg.
Spezifische Reaktivität Human
Konjugat Unconjugated
Kreuzreaktivität Hu
Spezies Human
Rekombinant Recombinant
Form Liquid
Protein-Subtyp Recombinant
Reinheits- oder Qualitätsgrad Affinity chromatography
Mehr anzeigen Weniger anzeigen

For Research Use Only.

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt