Learn More
Human Thyroglobulin Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Marke:  R&D Systems MAB8306-SP
Artikelnummer. 18703379
  • 201.00 € / 25 Mikrogramm
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Thyroglobulin Monoclonal specifically detects Thyroglobulin in Human samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
Thyroglobulin
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
AITD3, AITD3TGN, cog, TDH3, Tg, TGN, thyroglobulin
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
7038
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG1
Western Blot, Immunohistochemistry
904802
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL
P01266
TG
Chinese hamster ovary cell line CHO-derived recombinant human Thyroglobulin Met1-Lys2768 Accession # P01266
25 μg
Primary
Detects human Thyroglobulin in direct ELISAs.
Human
Purified
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen