Learn More
R&D Systems™ Human Tiki2/TRABD2B (NP_001181915) VersaClone cDNA
Beschreibung
This shuttle vector contains the complete ORF for the gene of interest, along with a Kozak consensus sequence for optimal translation initiation. It is inserted NotI to AscI. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector.
Bestellinformation
Ships at ambient temperature
Spezifikation
Spezifikation
| Gen | Tiki2/TRABD2B |
| Zugriffsnummer | NP_001181915 |
| Konzentration | 10 μg at 0.2 μg/μL |
| Zusammensetzung | cDNA is provided in 10mM Tris-Cl, pH 8.273 |
| Einsatztyp | 1567bp |
| Gen-ID (Entrez) | 388630 |
| Gen-Alias | HKAT, TIKI2, TRABD2B, TRABD2B TraB domain containing 2 B |
| Inhalt und Lagerung | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Spezies | Human |
| Menge | 10 μg |
For Reasearch Use Only. Not for use in Humans.