Learn More
Human TNF-alpha Antibody, R&D Systems™
GREENER_CHOICE
Artikelnummer. 18706019
Change view
Click to view available options
Menge:
100 μg
25 μg
Packungsgröße:
100 Mikrogramm
25 Mikrogramm
Artikelnummer. Menge unitSize
18706019 100 μg 100 Mikrogramm
18789128 25 μg 25 Mikrogramm
2 options
Artikelnummer. 18706019
Marke: R&D Systems MAB2102100
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Mouse Monoclonal Antibody
TNF-alpha Monoclonal specifically detects TNF-alpha in Human samples. It is validated for Western Blot, Immunocytochemistry.
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen TNF-alpha
Anwendungen Immunocytochemistry, Western Blot
Klassifikation Monoclonal
Klon 983003
Konjugat Unconjugated
Verdünnung Western Blot 1μg/mL, Immunocytochemistry 8-25μg/mL
Zusammensetzung Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS.
Gen-Zugriffsnummer P01375
Gen-Alias APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha
Wirtsspezies Mouse
Immunogen E. coli-derived recombinant human TNF-alpha Lys87-Ala232 Accession # P01375
Reinigungsverfahren Protein A or G purified from ascites
Menge 100 μg
Primär oder sekundär Primary
Gen-ID (Entrez) 7124
Testspezifität Detects human TNF-alpha in direct ELISAs.
Rekonstitution Reconstitute at 0.5mg/mL in sterile PBS.
Zielspezies Human
Inhalt und Lagerung Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b