missing translation for 'onlineSavingsMsg'
Learn More

Human TNF-alpha Antibody, R&D Systems™
GREENER_CHOICE

Artikelnummer. p-7139786
Change view
Click to view available options
Menge:
100 μg
25 μg
Packungsgröße:
100 Mikrogramm
25 Mikrogramm
Artikelnummer. Menge unitSize
18789128 25 μg 25 Mikrogramm
18706019 100 μg 100 Mikrogramm
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 18789128

Marke: R&D Systems MAB2102SP

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Mouse Monoclonal Antibody

TNF-alpha Monoclonal specifically detects TNF-alpha in Human samples. It is validated for Western Blot, Immunocytochemistry.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen TNF-alpha
Anwendungen Immunocytochemistry, Western Blot
Klassifikation Monoclonal
Klon 983003
Konjugat Unconjugated
Verdünnung Western Blot 1μg/mL, Immunocytochemistry 8-25μg/mL
Zusammensetzung Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS.
Gen-Zugriffsnummer P01375
Gen-Alias APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha
Wirtsspezies Mouse
Immunogen E. coli-derived recombinant human TNF-alpha Lys87-Ala232 Accession # P01375
Reinigungsverfahren Protein A or G purified from ascites
Menge 25 μg
Primär oder sekundär Primary
Gen-ID (Entrez) 7124
Testspezifität Detects human TNF-alpha in direct ELISAs.
Rekonstitution Reconstitute at 0.5mg/mL in sterile PBS.
Zielspezies Human
Inhalt und Lagerung Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Mehr anzeigen Weniger anzeigen
Name des Produkts
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.