Learn More
Human Transglutaminase 3/TGM3 Antibody, R&D Systems™
Sheep Polyclonal Antibody
Marke: R&D Systems AF4604
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Transglutaminase 3/TGM3 Polyclonal specifically detects Transglutaminase 3/TGM3 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Spezifikation
| Transglutaminase 3/TGM3 | |
| Polyclonal | |
| Unconjugated | |
| NP_003236 | |
| TGM3 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Transglutaminase 3/TGM3, Ala2-Glu693, Accession # NP_003236 | |
| 100 μg | |
| Primary | |
| Detects human Transglutaminase 3/TGM3 in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| EC 2.3.2.13, MGC126250, protein-glutamine gamma-glutamyltransferase E, TG(E), TG3, TGase E, TGase-3, TgaseE, TGEMGC126249, TGM3, transglutaminase 3 (E polypeptide, protein-glutamine-gamma-glutamyltransferase), Transglutaminase E, transglutaminase-3 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 7053 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |