Learn More
Human VISTA/B7-H5/PD-1H Antibody, R&D Systems™
Beschreibung
VISTA/B7-H5/PD-1H Monoclonal specifically detects VISTA/B7-H5/PD-1H in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF-ready.
Spezifikation
Spezifikation
| Antigen | VISTA/B7-H5/PD-1H |
| Anwendungen | Flow Cytometry, Immunocytochemistry, CyTOF |
| Klassifikation | Monoclonal |
| Klon | 730804 |
| Konzentration | LYOPH |
| Konjugat | Unconjugated |
| Verdünnung | Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready |
| Zusammensetzung | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
| Gen-Zugriffsnummer | Q9H7M9 |
| Gen-Alias | 4632428N05Rik, B7H5, B7-H5, C10orf54, chromosome 10 open reading frame 54, Dies1, Gi24, PD1H, PD-1H, platelet receptor Gi24, PP2135, SISP1, stress induced secreted protein 1, VSIR |
| Mehr anzeigen |