Learn More
R&D Systems™ Hyaluronan Quantikine ELISA Kit
Artikelnummer. p-7223013
Click to view available options
Menge:
1 Packung (6 Tupfer)
Eine 96-Well-Platte
Packungsgröße:
1 Set
96 Stück
Artikelnummer. 18742244
Marke: R&D Systems™ DHYAL0
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Hyaluronan Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
The Hyaluronan Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies multi-species Hyaluronan in cell culture supernates (50 ul), tissue homogenates (50 ul), serum (25 ul), edta plasma (25 ul), heparin plasma (25 ul).
TRUSTED_SUSTAINABILITY
Spezifikation
Assay-Bereich 0.63 - 40ng/mL
Assay-Empfindlichkeit: 0.2ng/mL
Konjugat HRP
Probentyp Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Tissue Homogenates
Spezifität 35 kDa Hyaluronan in Human, Mouse, Rat, Canine, and Porcine Samples
Zur Verwendung mit (Anwendung) ELISA
Menge Eine 96-Well-Platte
Probenvolumen Cell Culture Supernates (50uL),Tissue Homogenates (50uL),Serum (25uL),EDTA Plasma (25uL),Heparin Plasma (25uL)
Lagerungsbedingungen Store the unopened product at 2° to 8°C. Do not use past expiration date.