missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroxyzinpamoat, TRC
Specifications
Specifications
| Chemischer Name oder Material | Hydroxyzine Pamoate |
| CAS | 10246-75-0 |
| Summenformel | C23 H16 O6 . C21 H27 Cl N2 O2 |
| Menge | 5 g |
| Synonym | 2-Naphthalenecarboxylic acid, 4,4'-methylenebis[3-hydroxy-, compd. with 2-[2-[4-[(4-chlorophenyl)phenylmethyl]-1-piperazinyl]ethoxy]ethanol (1:1) (9CI, ACI), 2-Naphthoic acid, 4,4'-methylenebis[3-hydroxy-, compd. with 2-[2-[4-(p-chloro-α-phenylbenzyl)-1-piperazinyl]ethoxy]ethanol (1:1) (8CI), 2-Naphthoic acid, 4,4'-methylenebis[3-hydroxy-, hydroxyzine salt (6CI), Ethanol, 2-[2-[4-(p-chloro-α-phenylbenzyl)-1-piperazinyl]ethoxy]-, 4,4'-methylenebis[3-hydroxy-2-naphthoate] (1:1) (salt) (8CI), Ethanol, 2-[2-[4-[(4-chlorophenyl)phenylmethyl]-1-piperazinyl]ethoxy]-, 4,4'-methylenebis[3-hydroxy-2-naphthalenecarboxylate] (1:1) (9CI), 1-(p-Chloro-α-phenylbenzyl)-4-(2-hydroxyethoxyethyl)piperazine, pamoate, 2-(2-(4-((4-chlorophenyl)(phenyl)methyl)piperazin-1-yl)ethoxy)ethanol 4,4'-methylenebis(3-hydroxy-2-naphthoate), Atarax P, Equipose, Hydroxyzine pamoate, Hydroxyzyne pamoate, Masmoran, Paxisitil, Paxistil, Vistaril, Vistaril pamoate |
| InChI-Formel | InChI=1S/C23H16O6.C21H27ClN2O2/c24-20-16(14-7-3-1-5-12(14)9-18(20)22(26)27)11-17-15-8-4-2-6-13(15)10-19(21(17)25)23(28)29;22-20-8-6-19(7-9-20)21(18-4-2-1-3-5-18)24-12-10-23(11-13-24)14-16-26-17-15-25/h1-10,24-25H,11H2,(H,26,27)(H,28,29);1-9,21,25H,10-17H2 |
| SMILES | OCCOCCN1CCN(CC1)C(c2ccccc2)c3ccc(Cl)cc3.OC(=O)c4cc5ccccc5c(Cc6c(O)c(cc7ccccc67)C(=O)O)c4O |
| IUPAC-Name | 4-[(3-Carboxy-2-hydroxynaphthalin-1-yl)methyl]-3-hydroxynaphthalin-2-carbonsäure;2-[2-[4-[(4-Chlorphenyl)-phenylmethyl]piperazin-1-yl]ethoxy]ethanol |
| Molekulargewicht (g/mol) | 763.27 |
| Formelmasse | 762.2708 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?