Learn More
Ansell HyFlex™ 11581 Light Duty Touchscreen Compatible Cut-resistant Gloves
Artikelnummer. p-200019291 Alle ansehen Ansell Produkte
Change view
Click to view available options
Größe:
6
7
8
9
10
11
12
Packungsgröße:
12 Paare
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Größe unitSize
30282352 6 12 Paare
30282353 7 12 Paare
30282350 8 12 Paare
30282349 9 12 Paare
30282348 10 12 Paare
30282351 11 12 Paare
30282354 12 12 Paare
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Artikelnummer. 30282352 Lieferant Ansell Lieferanten-Nr. 11581060
Nur noch null auf Lager
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
HyFlex™ 11581 Light Duty Touchscreen Gloves are cut-resistant with a knit wrist cuff, making them ideal for industrial applications.
  • High cut protection with an immediate blade blunting effect outperforming cut protection of gloves in the same category
  • High dexterity and tactility
  • 21-gauge construction makes it the thinnest and lightest glove in its category
  • Touchscreen compatible, so it's ready to use in today's workplaces
  • Extended Use-FORTIX™ technology provides up to 20% more abrasion resistance for longer lasting handling in abrasive conditions
  • Reinforced thumb crotch
  • Made of tungsten, HPPE and nylon
  • Spandex seamless liner
  • DGX nitrile palm coating
TRUSTED_SUSTAINABILITY
Spezifikation
Coating Material Nitrile
Farbe Gray, Black
Cut Resistance Rating ANSI A6
Hand Specific
Latex Free Yes
Material Nylon
Produkttyp Cut-resistant Gloves
Größe 6