Learn More
Thermo Scientific™ HyperSep™ SPE Cartridges
Artikelnummer. 30279933 Alle ansehen Thermo Scientific Produkte
Ansicht ändern
Click to view available options
Sorbens:
HLB (Umkehrphasenpolymer)
Starker Anionenaustauscher im gemischten Modus, Polymer
Starker Kationenaustauscher im gemischten Modus, Polymer
Typ:
SPE-Patrone 10 mg/1 ml
SPE-Patrone 30 mg/1 ml
Packungsgröße:
100 Stück
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Sorbens Typ Packungsgröße
30279933 Starker Kationenaustauscher im gemischten Modus, Polymer SPE-Patrone 10 mg/1 ml 100 Stück
30279934 HLB (Umkehrphasenpolymer) SPE-Patrone 30 mg/1 ml 100 Stück
30279930 Starker Kationenaustauscher im gemischten Modus, Polymer SPE-Patrone 30 mg/1 ml 100 Stück
30279931 Starker Anionenaustauscher im gemischten Modus, Polymer SPE-Patrone 30 mg/1 ml 100 Stück
30279929 HLB (Umkehrphasenpolymer) SPE-Patrone 10 mg/1 ml 100 Stück
30279932 Starker Anionenaustauscher im gemischten Modus, Polymer SPE-Patrone 10 mg/1 ml 100 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Artikelnummer. 30279933 Lieferant Thermo Scientific™ Lieferanten-Nr. 60107101MCX
Verfügbar ab: 26-11-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
missing translation for 'validMainframeMsg'
Achieve greater reproducibility with cleaner more consistent extracts compared to conventional SPE, phospholipid removal and protein precipitation products.
Grow your laboratory productivity. Thermo Scientific™ HyperSep™ SPE Cartridges revolutionize solid phase extraction (SPE). This first fritless SPE product range enables greater sample throughput in bioanalytical laboratories where low elution volumes combined with greater reproducibility, lower failure rates and cleaner extracts are key to achieving better productivity.
HyperSep products are available in a 10 mg/1 mL cartridge format. They provide highly reproducible, clean and consistent sample extracts for bioanalysis by utilizing the unique award winning fritless SPE technology. The enhanced performance of HyperSep cartridges gives you greater confidence in your analytical results and lowers operating costs - without compromising ease of use or requiring complex method development.
HyperSep cartridges provide unparalleled performance characteristics compared to conventional SPE, phospholipid removal and protein precipitation products.
What HyperSep Cartridges Mean to Your Analysis
  • Higher levels of reproducibility
  • Higher levels of extract cleanliness
  • Reduced solvent requirements
  • Confidence in results
TRUSTED_SUSTAINABILITY
Spezifikation
Säulenbettgewicht 30 mg
Zur Verwendung mit (Anwendung) Bioanalytical Applications - Strong Ion-Exchange Retention of Weak Basic Compounds. Complementary Reversed Phase Retention of Neutral Compounds.
Produktlinie HyperSep
Sorbens Starker Kationenaustauscher im gemischten Modus, Polymer
Typ SPE-Patrone 10 mg/1 ml
Menge 100/pk